Orders LinkContactUsLinkComponent
Racks for test tubes
Racks for test tubes
24 mm, —, PE
Catalog # HACH1488600
Supplier:   Hach
Racks for test tubes
Catalog # HACH1488600
Supplier:  Hach
Racks for test tubes
Racks for test tubes
24 mm, —, PE
Price
25,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
These test tube racks features a special blend of polyethylene which sinks in water, assuring non-floating racks when immersed.
  • Stackable
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Hole Ø:
    24 mm
  • No. of Holes:
  • Material:
    PE
  • Seq:
    03