Order Entry
Belgium
Orders LinkContactUsLinkComponent
Carbonate Borax Free Buffer Solution

Carbonate Borax Free Buffer Solution

9,00, ±0,01, Carbonate, 20 °C, HDPE bottle, 1 L
Catalog # REGE1090BF
Supplier:   REAGECON
Carbonate Borax Free Buffer Solution
Catalog # REGE1090BF
Supplier:  REAGECON
CAS Number:  
Carbonate Borax Free Buffer Solution

9,00, ±0,01, Carbonate, 20 °C, HDPE bottle, 1 L

CAS Number:  
Price
62,70 €
Log in for availability
1 * 1 L
Product Details & Documents

This range of Reagecon pH Buffers are sodium tetraborate (Borax) free. They are not subject to classification by EU regulatory authorities as toxic to reproduction under CLP category 1B. Similar in approach to the existing range of Reagecon Mercury Free Buffers, these Buffer Solutions have been developed to enhance user choice and allay safety concerns, without any impact on fitness for purpose or product performance.

  • Non toxic and borax free
  • Reagecon pH buffers are pre-programmed into the instruments of most major manufacturers
  • Laboratory chemical

For increased confidence in their test measurements, analysts should regularly measure the pH of a control standard. If an acceptable value is obtained from the control standard measurement then the analysts can have improved confidence that their test measurements will be correct. Reagecon’s extensive range of pH Buffers means that there will be a Reagecon pH Buffer which can be used as a control buffer for all pH applications.

Manufactured to exacting specifications with an extended shelf life and stability, even after opening the bottle thereby eliminating the requirement to open a fresh bottle of solution every time it is used.

Certifications: Reagecon’s Borax free pH buffer solutions are tested by our ISO 17025 accredited test method and NIST traceable.

Packaging: HDPE bottles.

Specifications

Specifications

  • Pack type:
    HDPE bottle
  • pH buffer type:
    Carbonate
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • pH accuracy:
    ±0,01
  • pH value:
    9,00
  • Cat. No.:
    REGE1090BF
  • Physical form:
    Liquid

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.