GXP SOFTWARE BUNDLE PACKAGE 1 * 1 items
Catalog # MLDVSMP7GXPSTARTER
Supplier: Molecular Devices
Frequently Bought Together
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
ThermoMixer® C Thermomixer
Supplier: EPPENDORF
VWR® Counter-Pen™ Cell Counter
Supplier: VWR Collection
Titration beakers
Supplier: Mettler - Toledo
Accessory for airflow indicator
Supplier: Dräger
Accessories for Mettler balances
Supplier: Mettler - Toledo
SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: EPPENDORF
Gas detection tubes for hydrogen peroxide
Supplier: Dräger
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
pH/mV/°C meters, bench, FiveEasy™ F20 / FiveEasy™ Plus FP20
Supplier: Mettler - Toledo
Plastic cuvette for LICO
Supplier: Hach
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Consumables for infrared spectroscopy
Supplier: PerkinElmer
Accessories for titrators
Supplier: Mettler - Toledo
Melting point and boiling point capillaries and tubes
Supplier: BUCHI