CTLA4 MONOCLONAL ANTIBODY (1E6) 1 * 100 µG
Catalog # ENZOENZABS6960100
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
Eppendorf Tubes® Microtubes with Screw Cap, 5,0 ml
Supplier: EPPENDORF
Microplates, 96-well
Supplier: Greiner Bio-One
MicroWell™ Plates, 96 well, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Sample Dippers
Supplier: VWR Collection
Cell strainers, Falcon®
Supplier: Corning
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Polyester Films for ELISA and General Incubation
Supplier: VWR Collection
VWR® Bottles, Water Sampling
Supplier: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Culture media bottles, square, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
Finned plugs for tubes
Supplier: KARTELL