UBCH8 (HUMAN), (RECOMBINANT) (HIS-TAG) 1 * 100 µG
Catalog # ENZOBMLUW91350100
Supplier: ENZO LIFE SCIENCES
UBCH8 (HUMAN), (RECOMBINANT) (HIS-TAG) 1 * 100 µG
Catalog # ENZOBMLUW91350100
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
Serological pipettes
Supplier: Greiner Bio-One
VWR®, Flat Screw Caps for Microcentrifuge Tubes
Supplier: VWR Collection
Optical bottom plates, 96-well, Nunclon™Δ
Supplier: Thermo Fisher Scientific
Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Supplier: Corning
PCR plates, 96-well, low profile, half skirt, Axygen®
Supplier: Corning
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Roller bottle standard, surface treated, 1000 ml vent cap 1 * 24 items
Supplier: GUANGZHOU JET BIOFILTRATION
FalconRegistered Mark Round-Bottom Tubes, 14 mL 1 * 500 items
Supplier: STEMCELL Technologies
Stoppers, Pierce-It™
Supplier: Simport Scientific
Microcentrifuge tubes, 0,5/1,5/2,0 ml, with screw caps, Axygen®
Supplier: Corning
Multidishes, Nunc™ UpCell™ surface
Supplier: Thermo Fisher Scientific
Corning® Gosselin™ Spreaders
Supplier: Corning
FalconRegistered Mark Conical Tubes, 50 mL 1 * 500 items
Supplier: STEMCELL Technologies
Gel blotting sheets, GB005, Whatman™
Supplier: Whatman products (Cytiva)
Culture tubes, sterile, Cultubes™
Supplier: Simport Scientific