Orders LinkContactUsLinkComponent
Phosphoramidon
Phosphoramidon
25 mg
Catalog # ENZOBMLPI1110025
Supplier:   Enzo Life Sciences
Phosphoramidon
Catalog # ENZOBMLPI1110025
Supplier:  Enzo Life Sciences
CAS Number:  
Phosphoramidon
Phosphoramidon
25 mg
CAS Number:  
Price
886,00 €
Log in for availability
1 * 25 mg
Product Details & Documents
Neprilysin inhibitor
Inhibits neutral endopeptidase 24,11 (NEP, neprilysin; EC 3.4.24.11) (Ki=2 nM), a cell surface zinc metalloprotease which cleaves bioactive peptides such as bradykinin, neurotensin, and enkephalin. Also inhibits neprilysin 2 (Ki~2 nM). Potent inhibitor of endothelin-converting enzyme (ECE) (IC50=0,68 μM).
Specifications
Specifications
  • Pk:
    25 mg
  • Cat. No.:
    ENZOBMLPI1110025
  • Protein/peptide name:
    Phosphoramidon