Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cilostamide ≥98% (by TLC)

Cilostamide ≥98% (by TLC)

5 mg
Catalog # ENZOBMLPD1250005
Supplier:   Enzo Life Sciences
Cilostamide ≥98% (by TLC)
Catalog # ENZOBMLPD1250005
Supplier:  Enzo Life Sciences
CAS Number:  68550-75-4
Cilostamide ≥98% (by TLC)
Cilostamide ≥98% (by TLC)

5 mg

CAS Number:  68550-75-4
Price
86,90 €
Log in for availability
1 * 5 mg
Product Details & Documents

PDE inhibitor

Phosphodiesterase PDE3 inhibitor (IC50=5 nM). Blocks PKB/Akt signaling pathway by inhibition of Akt-activated PDE3B. Reverses the established effects of leptin on food intake and body weight. Blocks the leptin-induced tyrosine phosphorylation of STAT3 at the hypothalamic level.

Synonyms:  N-Cyclohexyl-N-methyl-4-((2-oxo-1,2-dihydroquinolin-6-yl)oxy)butanamide
  • CAS Number:68550-75-4
  • Formula:C₂₀H₂₆N₂O₃
  • MDL Number:MFCD00673958
  • Molecular Weight:342.43 g/mol
Specifications

Specifications

  • Pk:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.