GLUCAGON LIKE PEPTIDE 1 (HUMAN) (1-36) AMIDE 1 * 100 µG
Catalog # ENZOBMLGP14370100
Supplier: ENZO LIFE SCIENCES
GLUCAGON LIKE PEPTIDE 1 (HUMAN) (1-36) AMIDE 1 * 100 µG
Catalog # ENZOBMLGP14370100
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
Serological pipettes, Stripette™
Supplier: Corning
96-Well clear polystyrene microplates, Corning®
Supplier: Corning
VWR® Blender Bags
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Cell strainers, Falcon®
Supplier: Corning
Cell culture storage bottles, square, PC, Corning®
Supplier: Corning
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Cell culture flasks with standard or filter caps, CELLSTAR®
Supplier: Greiner Bio-One
PCR tubes and caps, Axygen®
Supplier: Corning
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Aspirating pipettes, Falcon®
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Detectable scoops, SteriPlast®
Supplier: BURKLE