BLOCKING PEPTIDE FOR DISHEVELLED-2 PAB (PROD. NO. BML-DA4270) 1 * 100 µG
ENZOBMLDP94270100
BLOCKING PEPTIDE FOR DISHEVELLED-2 PAB (PROD. NO. BML-DA4270) 1 * 100 µG
ENZOBMLDP94270100
Frequently Bought Together
Sealing tapes, Nunc™
: Thermo Fisher Scientific
Screw cap trays for 2D barcoded storage tubes
: THERMO MATRIX TECHNOLOGIES
NMR CAPILLARIES 75 MM 1.7 MM 1 * 10 Pce
: MARKET SOURCE PART PROCESS
Filter plates, 96-well, AcroPrep™
: Cytiva (Formerly Pall Lab)
Seals for ALPS™ 50 V heat sealer
: THERMO MATRIX TECHNOLOGIES
Conical Tubes with Screw Caps
: EPPENDORF
Screw caps for T501 sample tubes
: Simport Scientific
AcroPrep™ 24-Well Receiver Plate
: Cytiva (Formerly Pall Lab)
NANOPLATE 26K 240-WELL 1 * 10 items
: QIAGEN
Cellulose stoppers, Steristoppers®
: HERENZ HEINZ
1mL 96-Well Collection Plate with Round Well Bottoms, with SiliGuard™ Low Bind Coating 1 * 20 items
: MARKET SOURCE PART PROCESS
VWR® Cryoboxes, Double Size, 133x257 mm
: VWR Collection