Orders LinkContactUsLinkComponent
Serotonergic ligand library, Screen-Well®
Serotonergic ligand library, Screen-Well®
Screen-Well® Serotonergic ligand library, 100 µl/well, 1 SET
Catalog # ENZOBML28130100
Supplier:   Enzo Life Sciences
Serotonergic ligand library, Screen-Well®
Catalog # ENZOBML28130100
Supplier:  Enzo Life Sciences
CAS Number:  
Serotonergic ligand library, Screen-Well®
Screen-Well® Serotonergic ligand library, 100 µl/well, 1 SET
CAS Number:  
Price
2 690,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
The Screen-Well® serotonergic ligands plate contains 79 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The serotonergic ligands plate is 1 plate of the larger neurotransmitter library which contains over 680 ligands.
Specifications
Specifications
  • Pk:
    1 SET
  • Description:
    Screen-Well® Serotonergic ligand library, 100 µl/well