MULTIVIEW® Microarray Slides
Universal tissue microarray slides, 25 slides
Catalog # ENZOADI9500200025
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
These microarray slides with eleven human tissue specimens includes placenta, liver, tonsil, colon, skin, brain, breast (fibroadenoma), prostate, thyroid, kidney and fallopian tube.
- High-consistency specimen and slide preparation for reduced maintenance
- Fixed in formalin and paraffin embedded
- Sections are placed on a SuperFrost Plus slide
- Cores are approximately 2 mm in diameter
- Tissue sections are 3 µm thick
Suitable for use with common FFPE tissue protocols as rapid screening tools for the optimisation or validation of stains in a broad-spectrum of immunohistochemical procedures.
Specifications
- Description:Universal tissue microarray slides, 25 slides
- Seq:04
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



