Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Polysorbate 20, liquid

100 mL
Catalog # 1.37172.0100
Polysorbate 20, liquid
Catalog # 1.37172.0100
CAS Number:  9005-64-5
Polysorbate 20, liquid

100 mL

CAS Number:  9005-64-5
Price
Inquire For Price
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C
  • Melting Pt:98,9 °C
  • Density:1,1 g/cm³ (25 °C)
  • Flash Pt:275 °C
Specifications

Specifications

  • Pk:
    100 mL

Specifications