Height Adjustable Workstations
Product Details & Documents
The cleanroom height adjustable workstation is designed with motorised lifters to allow the operator to quickly change the height of the table to ensure a comfortable, ergonomic work environment.
Table top and leveler base are manufactured from stainless steel 304 with electro-polished finish. The adjustable legs are manufactured from anodised aluminum. Height adjustability is controlled from easy to use front mounted control unit.
Table height can be adjusted from 760 to 1147 mm (30 to 45"). Custom dimensions are possible.
These tables are designed for 220 V.
Specifications
- Description:Cleanroom height adjustable workstation, 1400×600×767-1147 mm
- Material:Stainless steel 304
- Seq:03
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



