ContactUsLinkComponent
,,5 components: S 2000mg/l P 500mg/l Sn 500mg/l Ti 500mg/l Si 100mg/l in HNO3 5% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM009338L1
,,5 components: S 2000mg/l P 500mg/l Sn 500mg/l Ti 500mg/l Si 100mg/l in HNO3 5% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM009338L1
Frequently Bought Together