TRYPAN BLUE 110ML 1 * 110 mL
Catalog # CVAT240102
Supplier: CELL VIABILITY
Frequently Bought Together
Potassium hydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Hydrogen peroxide 6% (w/v) (20 volumes), GPR RECTAPUR®
Supplier: VWR Chemicals
Spectroquant® PhotoCheck, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hypochlorite solution 5%, J.T.Baker®
Supplier: Avantor
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach