Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Polysorbate 20, liquid

Septum bottle, 1 L
Catalog # 1.37172.1000
Polysorbate 20, liquid
Catalog # 1.37172.1000
CAS Number:  9005-64-5
Polysorbate 20, liquid

Septum bottle, 1 L

CAS Number:  9005-64-5
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C
  • Melting Pt:98,9 °C
  • Density:1,1 g/cm³ (25 °C)
  • Flash Pt:275 °C
Specifications

Specifications

  • Pack type:
    Septum bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.