Order Entry
Belgium
Orders LinkContactUsLinkComponent
Polysorbate 80

Polysorbate 80

Septum bottle, 1 L
Catalog # 1.37171.1000
Polysorbate 80
Catalog # 1.37171.1000
CAS Number:  9005-65-6
Polysorbate 80

Septum bottle, 1 L

CAS Number:  9005-65-6
Price
3 910,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Polysorbate 80 high purity E can be used for pharmaceutical production and analysis.

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.

As part of our Emprove® program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Polysorbate 80 is used to stabilise biomolecule formulations and as solubiliser, emulsifier and wetting agent in small molecule formulations. Polysorbates are known to degrade via autoxidation and hydrolysis. Degradation products can have a negative impact on the stability of the protein as well as the small molecule API. To reduce this risk, the polysorbate high-purity grade contains a low amount of impurities, especially peroxides.

In addition, the product is protected from oxidation by the packaging system. The bottle contains a nitrogen overlay. A special lid allows removal of the content with a syringe without opening the bottle. Intended for high-risk applications, this Emprove® Expert grade contains bioburden and endotoxin limits.

Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Density:1,06…1,09 g/cm³ (25 °C)
  •  :M-Clarity™ quality level = MQ500 (more info.)
  • Flash Pt:>113 °C

Documents

Specifications

Specifications

  • Pack type:
    Septum bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.