Order Entry
Belgium
Orders LinkContactUsLinkComponent
WHEATON® Serum Vials

WHEATON® Serum Vials

int.Ø: 13 mm, ext.Ø: 20 mm (Mouth), 5,00 ml, Flat bottom, 21,75×40 mm, Glass, Amber, No
Catalog # WHEAWUBB5MLA-T-EP
Supplier:   DWK Life Sciences
WHEATON® Serum Vials
Catalog # WHEAWUBB5MLA-T-EP
Supplier:  DWK Life Sciences
WHEATON® Serum Vials
WHEATON® Serum Vials

int.Ø: 13 mm, ext.Ø: 20 mm (Mouth), 5,00 ml, Flat bottom, 21,75×40 mm, Glass, Amber, No

Price
113,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

These clear glass vials are manufactured from Wheaton '200' low extractable borosilicate glass to maintain uniform wall thickness. The amber vials are manufactured from Wheaton '320'  borosilicate glass.

  • Blowback feature in the neck and a specially designed bottom radius for added strength
  • These features are important for freeze-drying applications
  • Vials are provided in shrink-wrapped modules which greatly reduces particulate contamination

Certifications: Meets ASTM Type 1, Class A/B and USP Type 1 standards.

Delivery information: Vials are supplied without closures.

Specifications
  • Cap included:
    No
  • Ø×H:
    21,75×40 mm
  • Capacity:
    5,00 ml
  • Colour:
    Amber
  • Description:
    int.Ø: 13 mm, ext.Ø: 20 mm (Mouth)
  • Material:
    Glass
  • Seq:
    08
  • Base:
    Flat bottom
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.