VWR® Filter Vials
PES (Polyethersulfone), 0,45 µm
548-9049A
Filter vials with membrane, PE snap ring cap (11 mm), with hole and silicone white/PTFE red septa.
Vials have different colour caps for different membranes: RC with grey caps, PVDF with yellow caps, Nylon with light-blue caps, PTFE with pink caps and PES with light-green caps.
- Membrane:PES (Polyethersulfone)
- Pore size:0,45 µm
- Seq:04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



