Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

VWR® Filter Vials

PVDF (Polyvinylidene fluoride), 0,45 µm
  548-9043A
VWR® Filter Vials
  548-9043A
VWR® Filter Vials

PVDF (Polyvinylidene fluoride), 0,45 µm

 
247,00 €
   
1 * 100 ST
 

Filter vials with membrane, PE snap ring cap (11 mm), with hole and silicone white/PTFE red septa.

Vials have different colour caps for different membranes: RC with grey caps, PVDF with yellow caps, Nylon with light-blue caps, PTFE with pink caps and PES with light-green caps.

 
  • Membrane:
    PVDF (Polyvinylidene fluoride)
  • Pore size:
    0,45 µm
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.