Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Filter Vials

VWR® Filter Vials

RC (Regenerated cellulose), 0,45 µm
Catalog # 548-9041A
Supplier:   VWR Collection
VWR® Filter Vials
Catalog # 548-9041A
Supplier:  VWR Collection
VWR® Filter Vials

RC (Regenerated cellulose), 0,45 µm

Price
298,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

Filter vials with membrane, PE snap ring cap (11 mm), with hole and silicone white/PTFE red septa.

Vials have different colour caps for different membranes: RC with grey caps, PVDF with yellow caps, Nylon with light-blue caps, PTFE with pink caps and PES with light-green caps.

Specifications
  • Membrane:
    RC (Regenerated cellulose)
  • Pore size:
    0,45 µm
  • Seq:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.