(2S)-2-({[(9H-FLUOREN-9-YL)METHOXY]CARBO 1 * 1.000 mg
Catalog # MSPPCSMB00137700
Supplier: MARKET SOURCE PART PROCESS
(2S)-2-({[(9H-FLUOREN-9-YL)METHOXY]CARBO 1 * 1.000 mg
Catalog # MSPPCSMB00137700
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol denatured 96% ≥96%, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
Trichloroacetic acid (TCA) for synthesis, Sigma-Aldrich®
Supplier: Merck
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Glycerine, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers), extra pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.6%, Laboratory Reagent
Supplier: Honeywell Chemicals
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific