Orders LinkContactUsLinkComponent
Pass through cabinets
Pass through cabinets
Pass through cabinet, with static dissipative window, sealing gasket and compression handle, 2, 700×600×700 mm
Catalog # PALBPT-706070MS
Supplier:   Palbam
Pass through cabinets
Catalog # PALBPT-706070MS
Supplier:  Palbam
Pass through cabinets
Pass through cabinets
Pass through cabinet, with static dissipative window, sealing gasket and compression handle, 2, 700×600×700 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Pass Through cabinet allows products and materials to be transferred between different rooms. Reduces unnecessary walking and entering/exiting cleanroom.
  • Single or double versions available
  • Sealing gaskets and compression handles ensure firm closure
  • Tempered Glass or anti static PVC windows available
  • Mechanical or electronic interlocks available
  • Can be customised to requirements
Fabrication material: 304 stainless steel with electropolished finish.
Specifications
  • Description:
    Pass through cabinet, with static dissipative window, sealing gasket and compression handle
  • Seq:
    02
  • W×D×H:
    700×600×700 mm
  • Doors:
    2