Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

2-Methylindoline

Bottle, 25 g
Catalog # APOSOR10176-25G
Irritant
Supplier:   Apollo Scientific
2-Methylindoline
Catalog # APOSOR10176-25G
Supplier:  Apollo Scientific
CAS Number:  6872-06-6
2-Methylindoline

Bottle, 25 g

CAS Number:  6872-06-6
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:6872-06-6
  • Formula:C₉H₁₁N
  • MDL Number:MFCD00005706
  • Molecular Weight:133.19 g/mol
  • Boiling Pt:228…229 °C
  • Melting Pt:–51 °C
  • Flash Pt:93 °C
  • Storage Temperature:Ambient
  • EINECS:229-971-0
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications