Nile red
Bottle, 250 mg
Catalog # APOSBIN0465-250MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
- CAS Number:7385-67-3
- Formula:C₂₀H₁₈N₂O₂
- MDL Number:MFCD00011639
- Molecular Weight:318.38 g/mol
- Melting Pt:203…205 °C
- Storage Temperature:Ambient
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:250 mg



