ContactUsLinkComponent
7 components: Ca 40mg/l Cu 1mg/l Fe 400mg/l K 1400mg/l Mg 30mg/l P 300mg/l Zn 6mg/l in HNO3 5% 1 * 100 mL
Catalog # CMPLRM105963L1
7 components: Ca 40mg/l Cu 1mg/l Fe 400mg/l K 1400mg/l Mg 30mg/l P 300mg/l Zn 6mg/l in HNO3 5% 1 * 100 mL
Catalog # CMPLRM105963L1
Frequently Bought Together