Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Assistent® Brushes for Bottles

Nylon, 30 mm, Steel wire, 270 mm
  HECH91231200030
Assistent® Brushes for Bottles
  HECH91231200030
Assistent® Brushes for Bottles
Assistent® Brushes for Bottles

Nylon, 30 mm, Steel wire, 270 mm

 
48,10 €
   
1 * 10 ST
 

An assortment of brushes, available with nylon or natural bristles, designed for use with laboratory bottles and flasks.

  • For steep-bottom, round-shoulder, and threaded bottles and flasks

Brushes feature a head bundle for better cleaning of the bottoms of bottles and flasks.

 
  • Bristle material:
    Nylon
  • Brush head Ø:
    30 mm
  • Handle material:
    Steel wire
  • Seq:
    1
  • Total length:
    270 mm