Order Entry
Belgium
Orders LinkContactUsLinkComponent
Assistent® Brushes for Bottles

Assistent® Brushes for Bottles

Natural bristle, 15 mm, Steel wire, 280 mm
  HECH91120000015
Assistent® Brushes for Bottles
  HECH91120000015
Assistent® Brushes for Bottles
Assistent® Brushes for Bottles

Natural bristle, 15 mm, Steel wire, 280 mm

 
42,70 €
   
1 * 10 ST
 

An assortment of brushes, available with nylon or natural bristles, designed for use with laboratory bottles and flasks.

  • For steep-bottom, round-shoulder, and threaded bottles and flasks

Brushes feature a head bundle for better cleaning of the bottoms of bottles and flasks.

 
  • Bristle material:
    Natural bristle
  • Brush head Ø:
    15 mm
  • Handle material:
    Steel wire
  • Seq:
    5
  • Total length:
    280 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.