R-Phycoerythrin
Plastic bottle, 1 mg
Catalog # AATB2558
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
R-Phycoerythrin (PE) is isolated from red algae.
- CAS Number:11016-17-4
- MDL Number:MFCD00081950
- Molecular Weight:240 kDa
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:1 mg



