Order Entry
Belgium
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

Plastic bottle, 100 mL
Catalog # AATB11012
Irritant
Supplier:   AAT Bioquest
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)
Catalog # AATB11012
Supplier:  AAT Bioquest
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

Plastic bottle, 100 mL

CAS Number:  54827-17-7
Price
122,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Horseradish peroxidase (HRP) and HRP conjugates facilitate the ABTS oxidation in the presence of hydrogen peroxide, turning ABTS into its blue-green oxidized product.

  • Easy colorimetric detection
  • Easily measured via spectrophotometer
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL

Specifications