Order Entry
Belgium
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

Plastic bottle, 1 L
Catalog # AATB11003
Irritant
Supplier:   AAT Bioquest
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)
Catalog # AATB11003
Supplier:  AAT Bioquest
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)
3,3',5,5'-Tetramethylbenzidine (TMB) substrate, ReadiUse™ (colorimetric)

Plastic bottle, 1 L

CAS Number:  54827-17-7
Price
785,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Horseradish peroxidase (HRP) and HRP conjugates facilitate the ABTS oxidation in the presence of hydrogen peroxide, turning ABTS into its blue-green oxidized product.

  • Easy colorimetric detection
  • Easily measured via spectrophotometer
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.