Order Entry
Belgium
Orders LinkContactUsLinkComponent
Streptavidin

Streptavidin

Vial, 10 mg
Catalog # AATB16886
Supplier:   AAT Bioquest
Streptavidin
Catalog # AATB16886
Supplier:  AAT Bioquest
CAS Number:  
Streptavidin
Streptavidin

Vial, 10 mg

CAS Number:  
Price
785,00 €
Log in for availability
1 * 10 mg
Product Details & Documents

Streptavidin is a biotin-binding protein found in the culture broth of the bacterium Streptomyces avidinii.

  • Extremely high biotin-binding affinity
  • High quality

Caution: For research use only.

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Cat. No.:
    AATB16886
  • Protein/peptide type:
    Native
  • Protein/peptide name:
    Streptavidin
  • Protein synonyms:
    Avidin, Streptomyces avidinii
  • Conjugation:
    Unconjugated
  • Species:
    Streptomyces avidinii
  • Source:
    Streptomyces avidinii

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.