Orders LinkContactUsLinkComponent
PD-10 Spin Adapter
PD-10 Spin Adapter
PD-10 Spin adapter, 10 Pieces
Catalog # AATB60507
Supplier:   AAT Bioquest
PD-10 Spin Adapter
Catalog # AATB60507
Supplier:  AAT Bioquest
PD-10 Spin Adapter
PD-10 Spin Adapter
PD-10 Spin adapter, 10 Pieces
Price
142,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
PD-10 Spin adapters are convenient and rapid cleanup tools with high reproducibility and capacity for high throughput by running multiple samples in parallel with high desalting capacity.
  • Permits parallel sample processing
  • Efficient and convenient
  • Improves reproducibility
Caution: For research use only.
Specifications
  • Description:
    PD-10 Spin adapter, 10 Pieces
  • Seq:
    02