Order Entry
Belgium
Orders LinkContactUsLinkComponent
LDS 751 ≥95%

LDS 751 ≥95%

Plastic bottle, 25 mg
Catalog # AATB17561
Irritant
Supplier:   AAT Bioquest
LDS 751 ≥95%
Catalog # AATB17561
Supplier:  AAT Bioquest
CAS Number:  181885-68-7
LDS 751 ≥95%
LDS 751 ≥95%

Plastic bottle, 25 mg

CAS Number:  181885-68-7
Price
580,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

LDS 751 is a fluorescent cell-permeant nucleic acid stain that can be well excited with 488 nm laser line although it has a peak excitation at ~543 nm on dsDNA.

Synonyms:  LDS-751, 6-(Dimethylamino)-2-(4-(4-(dimethylamino)phenyl)buta-1,3-dien-1-yl)-1-ethylquinolin-1-ium perchlorate
  • CAS Number:181885-68-7
  • Formula:C₂₅H₃₀N₃·ClO₄
  • MDL Number:MFCD00467069
  • Molecular Weight:471.98 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.