Order Entry
Belgium
Orders LinkContactUsLinkComponent
Stand Rods, without Thread

Stand Rods, without Thread

13 mm, 250 mm
Catalog # BOCH5180-250
Supplier:   BOCHEM
Stand Rods, without Thread
Catalog # BOCH5180-250
Supplier:  BOCHEM
Stand Rods, without Thread

13 mm, 250 mm

Price
22,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, 18/10 steel.

  • Stand rods for stand bases
  • Without thread
Specifications
  • Ø:
    13 mm
  • Length:
    250 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.