NETZL.VII-H05VV-F3G1.0-C13/2.5M SW 1 * 1 items
Catalog # ZEIS0000000411763
Supplier: ZEISS
Frequently Bought Together
Accessories for empty columns HiScale™
Supplier: Cytiva
Gas detection tubes for ammonia
Supplier: Dräger
Cuvettes and tubes for Picco, Multy, Nova and Prove
Supplier: Merck
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
ThermoMixer® C Thermomixer
Supplier: EPPENDORF
SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: EPPENDORF
VWR® HCO 304, Conductivity Meter, Handheld
Supplier: VWR Collection
VWR®, Portable Turbidity Meter
Supplier: VWR Collection
Titration beakers
Supplier: Mettler - Toledo
GC-FID Autosampler
Supplier: LUCIDITY
GLASS CYLINDER FOR 10 ML BURETTE 1 * 1 items
Supplier: Mettler - Toledo
Accessories for titrators
Supplier: Mettler - Toledo
Storage phosphor screens and cassettes
Supplier: Cytiva
Melting point and boiling point capillaries and tubes
Supplier: BUCHI