1 COMPONENT: SILICATES 100 MG/L IN WATER 1 * 100 mL
Catalog # CMPLRM009184L1
Supplier: CUSTOM MADE CHEMICALS LAB
1 COMPONENT: SILICATES 100 MG/L IN WATER 1 * 100 mL
Catalog # CMPLRM009184L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Test kits, chromate, Spectroquant®, Supelco®
Supplier: Merck
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Soda lime, AnalaR NORMAPUR® analytical reagent, with indicator
Supplier: VWR Chemicals
pH indicator test strips, pH-Fix
Supplier: MACHEREY-NAGEL
Visual colour kits, copper, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier: VWR Chemicals
Immersion oil for microscopy, Sigma-Aldrich®
Supplier: Merck
Test strips, lactic acid, Reflectoquant®, Supelco®
Supplier: Merck
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hypochlorite solution 5%, J.T.Baker®
Supplier: Avantor
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
Bromocresol green-methyl red solution mixed indicator, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Acid red 52, Technical Grade, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY