ContactUsLinkComponent
18 components: 10mg/l each of Ce Dy Er Eu Gd Ho La Lu Nd Pr Sm Tb Th Tm Yb U Sr Pb in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15 1 * 100 mL
Catalog # CMPLRM103642L1
18 components: 10mg/l each of Ce Dy Er Eu Gd Ho La Lu Nd Pr Sm Tb Th Tm Yb U Sr Pb in HNO3 5% CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15 1 * 100 mL
Catalog # CMPLRM103642L1
Frequently Bought Together