Order Entry
Belgium
Orders LinkContactUsLinkComponent
GAP Hazardous Material Work Stations

GAP Hazardous Material Work Stations

Hazardous material work station GAP.140.090, 900×750×1400 mm, 865×650×1105 mm, 113,20 kg
Catalog # MSGA40582901
Supplier:   asecos
GAP Hazardous Material Work Stations
Catalog # MSGA40582901
Supplier:  asecos
GAP Hazardous Material Work Stations

Hazardous material work station GAP.140.090, 900×750×1400 mm, 865×650×1105 mm, 113,20 kg

Price
6 770,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Hazardous material work station, GAP model with robust construction made of chemically resistant, anodised aluminium profiles.

  • Required occupational exposure limits are maintained
  • Avoidance of dangerous explosive atmosphere
  • Used for testing of the air equipment according to EN 14175 Part 3 (5.4.4)
  • Maximum user safety
  • Transparent side panes made from 5 mm (toughened safety glass)
  • Highly effective capturing of hazardous materials inside the work station through special fresh air curtain technology
  • Prepared for connection to the on-site exhaust air system and for electrical supply (230 V/50 Hz)
  • For mounting on existing working surfaces

Standard equipment consists of:
Melamine resin coated rear wall
Exhaust and fresh air monitoring with visual and audible alarm
Interior lighting aranged anti-glare (LED 4000 K neutral white 500 lux on the work surface)
Transparent side walls
Opaque, fold-away deflector
Potential-free switching contact 4-pole (max.400 V/25 A)
Power cord 3 m

Specifications
  • Weight:
    113,20 kg
  • W×D×H ext.:
    900×750×1400 mm
  • Description:
    Hazardous material work station GAP.140.090
  • Seq:
    04
  • W×D×H int.:
    865×650×1105 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.