Order Entry
Belgium
Orders LinkContactUsLinkComponent
GAP Hazardous Material Work Stations

GAP Hazardous Material Work Stations

Hazardous material work station GAP.110.090.060, 900×600×1100 mm, 865×500×855 mm, 92,20 kg
Catalog # MSGA40563901
Supplier:   asecos
GAP Hazardous Material Work Stations
Catalog # MSGA40563901
Supplier:  asecos
GAP Hazardous Material Work Stations

Hazardous material work station GAP.110.090.060, 900×600×1100 mm, 865×500×855 mm, 92,20 kg

Price
6 670,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Hazardous material work station, GAP model with robust construction made of chemically resistant, anodised aluminium profiles.

  • Required occupational exposure limits are maintained
  • Avoidance of dangerous explosive atmosphere
  • Used for testing of the air equipment according to EN 14175 Part 3 (5.4.4)
  • Maximum user safety
  • Transparent side panes made from 5 mm (toughened safety glass)
  • Highly effective capturing of hazardous materials inside the work station through special fresh air curtain technology
  • Prepared for connection to the on-site exhaust air system and for electrical supply (230 V/50 Hz)
  • For mounting on existing working surfaces

Standard equipment consists of:
Melamine resin coated rear wall
Exhaust and fresh air monitoring with visual and audible alarm
Interior lighting aranged anti-glare (LED 4000 K neutral white 500 lux on the work surface)
Transparent side walls
Opaque, fold-away deflector
Potential-free switching contact 4-pole (max.400 V/25 A)
Power cord 3 m

Specifications
  • Weight:
    92,20 kg
  • W×D×H ext.:
    900×600×1100 mm
  • Description:
    Hazardous material work station GAP.110.090.060
  • Seq:
    01
  • W×D×H int.:
    865×500×855 mm