Orders LinkContactUsLinkComponent
Cold trap, Type GKF
Cold trap, Type GKF
Cold trap, borosilicate glass 3.3, condensate capacity 250 ml, KF NW25 NS
Catalog # KGWA1702-NW25
Supplier:   KGW
Cold trap, Type GKF
Catalog # KGWA1702-NW25
Supplier:  KGW
Cold trap, Type GKF
Cold trap, borosilicate glass 3.3, condensate capacity 250 ml, KF NW25 NS
Price
1 610,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This cold trap is a complete version in which the dewar flask is bonded to the cold trap, because of its design, this version has got two free walls for condensation. It is therefore particularly suited for trapping a high level of condensate from humidity, or solvents. This cold trap is equipped with a valve for draining off the condensate. It has got viewing strips as standard so that the LN₂ coolant level can easily be monitored. The standard cold trap is provided with conical ground joint connections NS 29/32.
  • Good chemical and thermal resistance
  • Viewing stripes for observation of the condensation process
  • Condensate outlet with O-ring sealing
  • Protecting cover made of aluminium
Used for gas cleaning by condensation e.g. in vacuum systems.
With special joints: Small flange KF NW 25, olive and DN flange 15-25.
Delivery information: Supplied with a lid.
Specifications
  • Description:
    Cold trap, borosilicate glass 3.3, condensate capacity 250 ml
  • Seq:
    03
  • Joint:
    KF NW25 NS