WHEATON® Serum Vials
int.Ø: 7 mm, ext.Ø: 13 mm (Mouth), 2,00 ml, Flat bottom, 14,75×32 mm, Glass, Amber, No
Catalog # WHEAWUBB2MLC-T-EP
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
These clear glass vials are manufactured from Wheaton '200' low extractable borosilicate glass to maintain uniform wall thickness. The amber vials are manufactured from Wheaton '320' borosilicate glass.
- Blowback feature in the neck and a specially designed bottom radius for added strength
- These features are important for freeze-drying applications
- Vials are provided in shrink-wrapped modules which greatly reduces particulate contamination
Certifications: Meets ASTM Type 1, Class A/B and USP Type 1 standards.
Delivery information: Vials are supplied without closures.
Specifications
- Cap included:No
- Ø×H:14,75×32 mm
- Capacity:2,00 ml
- Colour:Amber
- Description:int.Ø: 7 mm, ext.Ø: 13 mm (Mouth)
- Material:Glass
- Seq:03
- Base:Flat bottom
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



