Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Sample Splitters

VWR® Sample Splitters

8 partitions of 50 mm, RT 50
Catalog # HAVE205322280
Supplier:   VWR Collection
VWR® Sample Splitters
Catalog # HAVE205322280
Supplier:  VWR Collection
VWR® Sample Splitters
VWR® Sample Splitters

8 partitions of 50 mm, RT 50

Price
790,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

For dividing sieved material into representative test sieve samples.

Type RT 6,3 and 12,5: Consists of 1 lacquered sheet steel rack, 300×250×270 mm, 1 stainless steel divider and 3 receivers which are tin sheet (1,75 litres) or laminated plastic (2 litres).

Type RT 25; 37,5; 50 and 75: Consists of 1 rack 620×260×420 mm, 1 divider and 3 receivers with grab handles (8 litres), all of galvanised steel.

Consists of 1 rack 620×260×420 mm, 1 divider and 3 receivers with grab handles (8 litres), all of galvanised steel

Documents

Specifications
  • Description:
    8 partitions of 50 mm
  • Seq:
    10
  • Type:
    RT 50
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Collecting bin for sample splitter, receiver made from tinned sheet, 1,75 L
    Collecting bin for sample splitter, receiver made from tinned sheet, 1,75 L
    HAVE205326882
    Log in for availability
    1 * 1 ST - 25,30 €

Product Information

  • Catalog #
    Availability
    Price
  • Collecting bin for sample splitter, receiver plastic laminated, 2,5 L
    Collecting bin for sample splitter, receiver plastic laminated, 2,5 L
    HAVE205326899
    Log in for availability
    1 * 1 ST - 70,20 €

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.