Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
This kit is used to detect RNase contamination in sample. 1 * 1 KIT
Catalog # ABNOKA6885.1KIT
Supplier:
Abnova
Expand image
This kit is used to detect RNase contamination in sample. 1 * 1 KIT
Catalog # ABNOKA6885.1KIT
Supplier:
Abnova
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection