Orders LinkContactUsLinkComponent
Codesoft Lite Software
Codesoft lite subscription online
  BRDY196181
 :   Brady
Codesoft Lite Software
  BRDY196181
 :  Brady
Codesoft Lite Software
Codesoft lite subscription online
441,00 €
1 * 1 ST
Codesoft Lite label design software can be used for asset tracking, barcode labelling, and inventory and inspection labelling applications.
  • Only available as a single user license
  • Limited to 1 thermal printer
Languages availble include Arabic (Egypt), Czech, Danish, Dutch, English, Finnish, French, German, Hebrew, Hungarian, Italian, Norwegian (Bokmål), Polish, Portuguese, Romanian, Russian, Spanish, Swedish, and Turkish.
Compatible with BBP11, BBP12, BBP72, PR Plus, IP Series, PAM3000, PAM3600, THT 76mm Core, and TLS-PC LINK printers.
  • Description:
    Codesoft lite subscription online
  • Seq:
    01