Order Entry
Belgium
Orders LinkContactUsLinkComponent
Code Corporation Barcode Reader Accessories

Code Corporation Barcode Reader Accessories

Cable, black, 1,8 m cable length, code readers
Catalog # BRDY176701
Supplier:   Brady
Code Corporation Barcode Reader Accessories
Catalog # BRDY176701
Supplier:  Brady
Code Corporation Barcode Reader Accessories
Code Corporation Barcode Reader Accessories

Cable, black, 1,8 m cable length, code readers

Price
17,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Accessories available for barcode readers.

Use the 1,8 m (BRDY176701) or 2,4 m (BRDY176700) coiled RS232 Affinity® cable CRA-C511 with code's cabled barcode readers.

Model BRDY176702 code power supply is an international power supply with adapter clips for the US, EU, UK and AU (barrel plug, 12 V/3,3 A).

Model BRDY176703 code power supply is an international power supply with USB connection.

Specifications
  • Description:
    Cable, black, 1,8 m cable length, code readers
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.