Orders LinkContactUsLinkComponent
Resolvex® Column Temperature Controller
Column cooler 250 CERA, 1,5×4,0×17"
  TCAT282-0300
 :   Tecan
Resolvex® Column Temperature Controller
  TCAT282-0300
 :  Tecan
Resolvex® Column Temperature Controller
Column cooler 250 CERA, 1,5×4,0×17"
1 * 1 ST
Column temperature controllers for heating and cooling.
  • Reliable PID microprocessor controlled thermal circuit stabilizes the operating environment
  • Easily-seated thermal blanket encloses the column in horizontal, vertical or slant orientation
  • Heater inserts are modular for easy removal and arrangement
Based on a unique, space-saving concept for efficient temperature control of liquid chromatography columns. Column enclosure accepts ³/₈" ext. Ø HPLC columns of any length up to 30 centimeters (15 centimeters in Column Heater 150).
  • Description:
    Column cooler 250 CERA, 1,5×4,0×17"
  • Seq:
    03