Resolvex® Column Temperature Controller
Column heater 150 CERA, 1,5×1,5×10"
Catalog # TCAT282-0240
Supplier: Tecan
Jump to:
Product Details & Documents
Column temperature controllers for heating and cooling.
- Reliable PID microprocessor controlled thermal circuit stabilizes the operating environment
- Easily-seated thermal blanket encloses the column in horizontal, vertical or slant orientation
- Heater inserts are modular for easy removal and arrangement
Based on a unique, space-saving concept for efficient temperature control of liquid chromatography columns. Column enclosure accepts ³/₈" ext. Ø HPLC columns of any length up to 30 centimeters (15 centimeters in Column Heater 150).
Specifications
- Désignation:Column heater 150 CERA, 1,5×1,5×10"
- Seq:01