Doypack® Paper/PET/CPP Paper Bags
Product Details & Documents
The specific shape of the Doypack® bags allows a good vertical position of the packaging on the shelves. The Paper/PET/CPP laminate provides a good barrier to gases and humidity. The paper layer gives the bag a natural look.
- Food contact
- Available in different sizes
- Ideal for wet and dry content
- Doypack® packaging
- Ideal for shelf display
- Available in stock
The Doypack® packaging has a window to give a better visibility of the content and highlights the features of the product. Ideal for food products (biscuits, confectionery, snacks) this bag is reclosable and reusable. All Doypack® Stand Up pouches in stock are equipped with a Clicky® closure system, with tactile & sound effects for an intuitive and sensory closure.
Specifications
- Capacity:500 ml
- Description:Doypack® Paper/PET/CPP bags, 2×35 mm bottom gusset
- Material:PET/CPP
- Seq:03
- Thickness:PAPER40/PET12/CPP60
- W×L:130×190 mm
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



