Order Entry
Belgium
Orders LinkContactUsLinkComponent
Doypack® Paper/PET/CPP Paper Bags

Doypack® Paper/PET/CPP Paper Bags

Doypack® Paper/PET/CPP bags, 2×32 mm bottom gusset, 250 ml, PET/CPP, PAPER40/PET12/CPP60, 110×160 mm
Catalog # 129-3902
Supplier:   Flexico
Doypack® Paper/PET/CPP Paper Bags
Catalog # 129-3902
Supplier:  Flexico
Doypack® Paper/PET/CPP Paper Bags

Doypack® Paper/PET/CPP bags, 2×32 mm bottom gusset, 250 ml, PET/CPP, PAPER40/PET12/CPP60, 110×160 mm

Price
350,00 €
Log in for availability
1 * 500 ST
Product Details & Documents

The specific shape of the Doypack® bags allows a good vertical position of the packaging on the shelves. The Paper/PET/CPP laminate provides a good barrier to gases and humidity. The paper layer gives the bag a natural look.

  • Food contact
  • Available in different sizes
  • Ideal for wet and dry content
  • Doypack® packaging
  • Ideal for shelf display
  • Available in stock

The Doypack® packaging has a window to give a better visibility of the content and highlights the features of the product. Ideal for food products (biscuits, confectionery, snacks) this bag is reclosable and reusable. All Doypack® Stand Up pouches in stock are equipped with a Clicky® closure system, with tactile & sound effects for an intuitive and sensory closure.

Specifications
  • Capacity:
    250 ml
  • Description:
    Doypack® Paper/PET/CPP bags, 2×32 mm bottom gusset
  • Material:
    PET/CPP
  • Seq:
    02
  • Thickness:
    PAPER40/PET12/CPP60
  • W×L:
    110×160 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.